missing translation for 'onlineSavingsMsg'
Learn More

Ansell VersaTouch™ 92-471 Series Light Blue Nitrile Gloves

Product Code. 12617071 Shop All Ansell Products
Click to view available options
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12617071

Brand: Ansell 92471100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Made from innovative nitrile compound which offers good chemical and mechanical properties, comfort and dexterity.

  • Textured finish on fingertips
  • Improved grip
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Outstanding resistance to animal fats compared with PVC or natural rubber
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves

Applications:

  • All final stages of meat or fish processing
  • Controlled-environment rooms
  • Prepared meals
  • Dairy processing
  • Catering
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Mark, Standard CATEGORY III EN 374
Product Type Controlled Environment Gloves
Sterility Non-sterile
Thickness (Metric) 0.12 mm
Quantity 100 Pairs
Resistant To Mechanical and chemical
Series 92-471
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.