missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Versatouch™ 72-287 Series Light Blue Dyneema™ Fiber Heavy Weight Gloves

Provides cut resistance, comfort and durability.

Brand:  Ansell Edmont™ 72287100

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12679761

  • £150.40 / Pack of 6


Estimated Shipment
14-02-2018
Add to basket

Description

Description

  • Contains Dyneema™
  • High cut resistance
  • Washable up to 90°C
  • Suitable for contact with all foodstuffs

Applications:

  • Boning
  • Carving
  • Fish processing
  • Fish filleting
  • Meals preparation
  • Fruit and vegetable processing
Specifications

Specifications

Dyneema® Fiber
Knitwrist
Dyneema® Fiber
72 Pairs
Heavy Weight Gloves
Dyneema® Fiber
Dyneema® Fiber
Cut resistant
CE Mark, Standard CATEGORY II EN 388 : 254X
Videos