missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Versatouch™ 72-287 Series Dyneema™ Fiber Heavy Weight Gloves

Catalog Number p-8005388
This item is not returnable. View return policy
This item is not returnable. View return policy

Excellent cut performance with ergonomic design to improve comfort. Suitable for contact with all foodstuffs.

  • Contains Dyneema™
  • High cut resistance
  • Washable up to 90°C
  • Suitable for contact with all foodstuffs

Applications:

  • Boning
  • Carving
  • Fish processing
  • Fish filleting
  • Meals preparation
  • Fruit and vegetable processing
Back Material Dyneema™ Fiber
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 254X
Cuff Material Dyneema™ Fiber
Product Type Heavyweight Gloves
Glove Cuff Style Knitwrist
Liner Material Dyneema™ Fiber
Lining Dyneema™ Fiber
Quantity 72 Pairs
Resistant To Cut resistant
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'