Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Filter
3M Versaflo™ TR-600 PAPR Accessory, Filter is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand:  3M™ 7100020996
Additional Details : Weight : 3.20000kg
Product Code. 15539295
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Simple to install
  • For use with  3M Versaflo Powered Air TR-600 series 
  • Recommends use of TR-6100FC filter cover to help protect filter from physical damage