Learn More
3M™ Versaflo™ TR-3712E P3 Particulate Filter
For use with 3M™ Versaflo™ Powered Air Respirator TR-300+. 3M™ High Efficiency P3 Particulate filter can be changed quickly and easily by hand.
Brand:  3M™ 7100018965
Code : NEW
Additional Details : Weight : 0.16200kg
Product Code. 15759067
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • High efficiency P3 particulate filter
  • Quick and easy changeover by hand
  • EN 12941,‎ CE Approved
  • For use with the 3M™ TR-300+ Powered Air Respirator series