Learn More
3M™ Versaflo™ Filter TR-3802E
3M™ Versaflo™ Filter TR-3802E for use with 3M™ Versaflo™ Powered Air Respirator TR-300+.
Brand: 3M™ 7100018809_OLD
Code : NEW
Additional Details : Weight : 0.19350kg
Description
- P3 particulate filter with nuisance level organic vapor relief
- The filter can be changed quickly and easily by hand
- For use 3M™ TR-300+ Powered Air Respirator
Specifications
CE Approved, EN 12941 | |
Filter | |
Nuisance level organic vapor relief | |
0.352 x 0.2 x 0.13 m |
P3 Particulate | |
3M™ Versaflo™ Powered Air Respirator TR-300+ | |
TR-3802E |