Learn More
3M™ Versaflo™ Comfort Pad for S-950 Premium Head Suspension
For use with S-950 Premium Head Suspension
Brand: 3M™ VMI7000042685
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- For use with S-950 Premium Head Suspension
- Wrapped around headband to create more comfortable fit and to absorb sweat
- 3M Catalog# S-953 , 3M Catalog# S-954