missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Veratrole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 21°C to 23°C |
Density | 1.084 |
Boiling Point | 206°C to 207°C |
Flash Point | 87°C (188°F) |
Refractive Index | 1.533 |
Quantity | 250 g |
Beilstein | 1364621 |
Merck Index | 14,9956 |
Solubility Information | Soluble in alcohol,diethyl ether,acetone,and methanol. Slightly soluble in water. |
Formula Weight | 138.17 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'