Learn More
Veratridine, Tocris Bioscience™
Voltage-gated Na+ channel opener
Brand:  Tocris (Bio-Techne) 291810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13462044
  • £103.95 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Voltage-gated Na+ channel opener; increases intracellular Ca2+ with no effect on the Na+/Ca2+ exchanger. Steroid-derived alkaloid neurotoxin. Increases membrane Na+ permeability and depolarizes excitable tissues.
Specifications
Chemical Identifiers
71-62-5
673.8
veratridine
CC1CCC2C(C3(C(CC4(C5CCC6C7(C5(CC4(C3CN2C1)O)OC6(C(CC7)OC(=O)C8=CC(=C(C=C8)OC)OC)O)C)O)O)O)(C)O
C36H51NO11
FVECELJHCSPHKY-IBETYFJASA-N
132991793
Specifications
Veratridine
71-62-5
veratridine
CC1CCC2C(C3(C(CC4(C5CCC6C7(C5(CC4(C3CN2C1)O)OC6(C(CC7)OC(=O)C8=CC(=C(C=C8)OC)OC)O)C)O)O)O)(C)O
132991793
>98%
10mg
C36H51NO11
FVECELJHCSPHKY-IBETYFJASA-N
673.8
673.79
Videos