Learn More
Thermo Scientific™ Veratridine, ≥97%
Activates sodium ion channels
Brand: Alfa Aesar™ J62324.MA
Code : NEW
Additional Details : CAS Number : 71-62-5 Weight : 0.00010kg Transport : UN number : 1544 Chem class : 6.1 Pack group : I
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
10 mg | |
-30°C to -10°C | |
≥97% | |
C36H51NO11 | |
3-Veratroylveracevine | |
Signal Transduction Reagent |
White | |
Activates sodium ion channels | |
71-62-5 | |
673.80g/mol | |
Soluble in ethanol or DMSO; Freely soluble in chloroform; Also soluble in 1M HCl | |
Powder |