Learn More
Emma Safety Footwear Vera XD Safety Shoes, Green
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and dirt resistant.
Brand: Emma Safety Footwear MM50750439
Additional Details : Weight : 1.27200kg
Description
- S2-Safety shoe for wet working conditions
- GripForce™ Easy Twist
- Microfiber construction
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
Vera XD, Green | |
Microfiber | |
39 |