Learn More
Veolia Water Systems™ Deionization Cartridge
Robust and reliable water purification cartridges ideal for removing ionic contaminants to achieve specified water quality.
Brand: Veolia Water Systems LC111
Code : A0
Additional Details : Weight : 2.42000kg
Description
- Deionization cartridges
- Removes ionic contaminants to achieve specified water quality
- Compatible with Maxima MK1/MK2
Specifications
Removes ionic contaminants to achieve specified water quality | |
Maxima MK1/MK2 |