Learn More
Emma Safety Footwear Venus S3 Safety Shoes
The Venus is a low profile, easy on / easy off shoe. The smooth, easy to clean contours offer additional hygiene advantages over and above lace up shoes. It's perfect for a 'quick change' from office to factory floor or walking between one facility and another.
Brand: Emma Safety Footwear MM71454647
Additional Details : Weight : 1.79200kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
47 |
Elastic | |
Venus S3 | |
Leather | |
Steel |