Learn More
Venlafaxine hydrochloride, Tocris Bioscience™
5-HT and NA reuptake inhibitor (SNRI)
Brand:  Tocris (Bio-Techne) 291710
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13442044
  • £114.75 / 10mg
Estimated Shipment
27-06-2019
Description
Description
Dual serotonin/noradrenalin re-uptake inhibitor that displays ∽ 30-fold higher affinity for SERT than NET (Ki values are 82 and 2480 nM respectively). Antidepressant; increases swimming and climbing behavior in the forced-swim test in rats.
Specifications
Chemical Identifiers
99300-78-4
313.866
venlafaxine hydrochloride, effexor, venlafaxine hcl, effexor xr, trevilor, dobupal, efexor, vandral, venlafaxinehydrochloride, venlafaxine hydrochloride
CHEBI:9944
CN(C)CC(C1=CC=C(C=C1)OC)C2(CCCCC2)O.Cl
C17H28ClNO2
QYRYFNHXARDNFZ-UHFFFAOYSA-N
62923
1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
Specifications
Venlafaxine hydrochloride
99300-78-4
venlafaxine hydrochloride, effexor, venlafaxine hcl, effexor xr, trevilor, dobupal, efexor, vandral, venlafaxinehydrochloride, venlafaxine hydrochloride
CN(C)CC(C1=CC=C(C=C1)OC)C2(CCCCC2)O.Cl
313.866
CHEBI:9944
>99%
10mg
C17H28ClNO2
QYRYFNHXARDNFZ-UHFFFAOYSA-N
1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
62923
313.86
Videos