missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Receptor 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111235
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: LEWAWPGAQE APATGDKDSE DTGVVRDCEG TDARPYCKVL LLHEVHANDT GSYVCYYKYI KARIEGTTAA SSYVFVRDFE QPFINKPDTL LVNRKDAMWV PCLVSIPGLN VTLRSQSSVL WPDGQEVVWD DRRGMLVSTP LLH
VEGF Receptor 3 Polyclonal antibody specifically detects VEGF Receptor 3 in Human samples. It is validated for Immunocytochemistry.Specifications
VEGF Receptor 3 | |
Polyclonal | |
Unconjugated | |
FLT4 | |
AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P35916 | |
FLT4 | |
Recombinant Protein corresonding to Human VEGF Receptor 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |