missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-D Rat anti-Mouse, Clone: 90409, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | VEGF-D |
---|---|
Clone | 90409 |
Applications | Western Blot, ELISA |
Conjugate | Unconjugated |
Form | Purified |
Description
VEGF-D Monoclonal specifically detects VEGF-D in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
VEGF-D | |
Western Blot, ELISA | |
Purified | |
RUO | |
2277 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
90409 | |
Unconjugated | |
Rat | |
Mouse | |
E. coli-derived recombinant mouse VEGF-D Phe98-Ser206 Accession # P97946 | |
Primary |
For Research Use Only