Learn More
VDM 11, Tocris Bioscience™
Potent, selective anandamide transport inhibitor
Brand:  Tocris (Bio-Techne) 139225
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 25mg
 View more versions of this product
Product Code. 13439293
  • £316.05 / 25mg
Estimated Shipment date
from Supplier 28-10-2019
Description
Description
A potent and selective inhibitor of the anandamide membrane transporter (AMT) with IC50 values of 4-11 μM. Displays negligible agonist activity at the hVR1 receptor and very weak action at CB1 and CB2 receptors. Ki values are > 5-10 μM at CB1 and CB2. Active in vivo. Also available in water-soluble emulsion (Cat. No. 1686).
Specifications
Chemical Identifiers
313998-81-1
409.614
vdm-11, n-4-hydroxy-2-methylphenyl arachidonylamide
(5E,8E,11E,14E)-N-(4-hydroxy-2-methylphenyl)icosa-5,8,11,14-tetraenamide
C27H39NO2
WUZWFRWVRHLXHZ-SHDWVJIKSA-N
71308297
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=C(C=C(C=C1)O)C
Specifications
25mg
313998-81-1
vdm-11, n-4-hydroxy-2-methylphenyl arachidonylamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=C(C=C(C=C1)O)C
409.614
409.61
VDM 11
C27H39NO2
WUZWFRWVRHLXHZ-SHDWVJIKSA-N
(5E,8E,11E,14E)-N-(4-hydroxy-2-methylphenyl)icosa-5,8,11,14-tetraenamide
71308297
>98%