Learn More
Invitrogen™ Human VCAM-1, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75752
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
VCAM1 (human) Recombinant Protein, RP-75752, from Invitrogen.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases. This product contains a purified 674 amino acid fragment (aa25-698) corresponding to the mature extracellular VCAM protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.Specifications
P19320 | |
tris HCl with 50% glycerol, 5mM EDTA and no preservative | |
Human VCAM-1 | |
E. Coli | |
RUO | |
VCAM1 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control, Western Blot | |
7412 | |
100μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human VCAM-1 | |
Human | |
Hu | |
Recombinant | |
Recombinant |