missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAT1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548513
Additional Details : Weight : 0.01000kg
Description
VAT1L Polyclonal antibody specifically detects VAT1L in Human, Mouse samples. It is validated for Western Blot.Specifications
VAT1L | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q80TB8, Q9HCJ6 | |
VAT1L | |
KLH conjugated synthetic peptide between 391-419 amino acids from the C-terminal region of human VAT1L. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
VAT1L | |
9430073I07; AI427515; Kiaa1576; mKIAA1576; Synaptic vesicle membrane protein VAT-1 homolog-like; Vat1l; vesicle amine transport 1-like; vesicle amine transport protein 1 homolog (T. californica)-like; vesicle amine transport protein 1 homolog-like (T. californica) | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
270097, 57687 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |