Learn More
Varenicline tartrate, Tocris Bioscience™
Selective α4β2 partial agonist; orally active
Brand:  Tocris (Bio-Techne) 375410
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13443554
  • £196.68 / 10mg
Estimated Shipment
09-07-2019
Description
Description
Orally active, subtype-selective partial agonist at α4β2 nicotinic receptors (Ki values are 0.06, 240, 322 and 3540 nM for α4β2, α3β4, α7, α1βγδ receptors respectively). Reduces nicotine-evoked dopamine release in vitro and decreases nicotine self-administration in vivo. Also potently activates PSAM4-5-HT3 and PSAM4-GlyR chimeric ion channels (EC50 values are 4 and 1.6 nM, respectively)
Specifications
Chemical Identifiers
375815-87-5
361.354
varenicline tartrate
C1C2CNCC1C3=CC4=NC=CN=C4C=C23.C(C(C(=O)O)O)(C(=O)O)O
C17H19N3O6
TWYFGYXQSYOKLK-ZBABXEQTSA-N
124080896
Specifications
Varenicline tartrate
375815-87-5
varenicline tartrate
C1C2CNCC1C3=CC4=NC=CN=C4C=C23.C(C(C(=O)O)O)(C(=O)O)O
124080896
>99%
10mg
C17H19N3O6
TWYFGYXQSYOKLK-ZBABXEQTSA-N
361.354
361.37
Videos