Learn More
Ansell Edmont™ Vantage™ 70-766 Series Green and Gray Kevlar™ Heavy Weight Gloves
Advanced cut protection combined with premium leather for superior grip.
Brand:  Ansell Edmont™ 70766100
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12676951
  • £298.67 / 12 pairs
Estimated Shipment
19-03-2018
Add to basket
Description
Description
  • Heavy duty gloves
  • With reinforced thumb and seams relocated to the backs of the fingers
  • Reduces exposure to cuts and offers greater protection and longer life
  • Helps guard against minor injuries and punctures
  • Offers superior grip even in oily environments
  • Longer cuff for additional protection
  • Plaited inside for comfort with coloured polyester/cotton
  • Palm reinforced with premium leather
Applications:
  • Press and stamping
  • Handling heavy and oily metal pieces
  • Handling sharp or irregular-edged metal parts
Advanced cut protection combined with premium leather for superior grip.
Specifications
Specifications
Kevlar®
Long knit wrist
Leather pad palm
High cut resistance
CE Mark, EN 388 : 4443, EN 407
Kevlar®
Kevlar®
Kevlar®
12 Pairs
Heavy Weight Gloves
Videos