missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadyl phthalocyanine, 85%, pract., Thermo Scientific Chemicals
141.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 80 % min |
Packaging | Glass bottle |
Quantity | 25 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in xylene,2-ethylhexanoic acid,,2-ethylhexylamine and 6-methyl-2,4-heptanedione |
Formula Weight | 579.46 |
Percent Purity | 85% |
Physical Form | Crystalline Powder, Crystals or Chunks |
Chemical Name or Material | Vanadyl phthalocyanine |
Safety and Handling
Product Identifier
- Vanadyl phthalocyanine
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'