missing translation for 'onlineSavingsMsg'
Learn More

Vanadium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog Number 10501953
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7803-55-6, 7697-37-2, 7732-18-5
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
516859
N.O[V](=O)=O
This item is not returnable. View return policy
7803-55-6, 7697-37-2, 7732-18-5
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
516859
N.O[V](=O)=O

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
V1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 1.0, 15.0, 84.0
CAS Max % 1.0, 15.0, 84.0
IUPAC Name vanadiumoylol amine
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2% HNO3    1000μg/mL    PLV2-2Y, -2X
15% HNO3    10,000    PLV4-3Y, -3X
2% HCl    1000μg/mL    PLV1-2X
15% HCl    10,000    PLV3-3X
Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up 30 elements at ppb levels.
Solvent or Matrix 2% HNO3
Quantity 500 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Vanadium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000 µg/mL Vanadium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, ammonium metavanadate

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'