missing translation for 'onlineSavingsMsg'
Learn More

Vanadium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11301599
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
This item is not returnable. View return policy
7440-62-2
V
50.94
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
23990
CHEBI:35170
vanadium
[V]
This item is not returnable. View return policy
7440-62-2
V
50.94
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
23990
CHEBI:35170
vanadium
[V]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Melting Point 1900°C
Boiling Point 3000°C
Appearance Silver Gray powder
Density 6.11
Assay Percent Range 99.5% (Metals basis)
Quantity 1 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Vanadium
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'