missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(IV) chloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 153.0°C |
Melting Point | -20.5°C |
Color | Brown-Red |
Physical Form | Liquid |
Quantity | 25 g |
Assay Percent Range | 99% min. (VCl4) |
Linear Formula | VCl4 |
Solubility Information | Solubility in water: vigorous reaction. Other solubilities: soluble in absolute alcohol, ether, chloroform,, acetic acid |
Formula Weight | 192.75 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Product Identifier
- Vanadium(IV) chloride
- Danger
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- EUH029-Contact with water liberates toxic gas.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'