missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Vanadium(IV) chloride, 99+%, 99.99% (metals basis)

Brand:  Alfa Aesar™ 087685.35

Code : NEW

Additional Details : CAS Number : 7632-51-1 Weight : 0.45000kg Transport : UN number : 2444 Chem class : 1a Pack group : I

Quantity 450g

Product Code. 15487505

  • £1324.15 / 450g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Specifications

Vanadium (IV) chloride
Liquid
(metals basis)
192.75
UN2444
1.816g/mL at 30°C
Moisture Sensitive
−28°C
7632-51-1
≥99%
VCl4
MFCD00061343
Vanadium tetrachloride
Cylinder with ball valve
148.5°C (755mmHg)
450g
Videos
SDS