missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals

Catalog Number 11314919
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
10049-12-4
F3V
107.94
MFCD00042553
PASVSMBJEYQYSN-UHFFFAOYSA-K
Vanadium trifluoride
vanadium(3+) trifluoride
[F-].[F-].[F-].[V+3]
This item is not returnable. View return policy
10049-12-4
F3V
107.94
MFCD00042553
PASVSMBJEYQYSN-UHFFFAOYSA-K
Vanadium trifluoride
vanadium(3+) trifluoride
[F-].[F-].[F-].[V+3]

Employed in oxygen-sensitive applications, such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ≥1406°C
Physical Form Powder
Quantity 2 g
UN Number UN2923
Merck Index 14,9922
Solubility Information Practically insoluble in water, acetone, and glacial acetic acid
Formula Weight 107.94
Percent Purity 98%
Sensitivity Moisture sensitive
Density 3.363 g/mL at 19°C
Chemical Name or Material Vanadium (III) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'