Learn More
Tocris Bioscience™ Valsartan
High affinity, selective AT1 antagonist
Brand:  Tocris Bioscience™ 421610
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13444374
  • £124.20 / 10mg
Estimated Shipment
28-06-2019
Description
Description
High affinity AT1 receptor antagonist (Ki = 2.38 nM). Displays 30,000-fold selectivity over AT2 receptors. Inhibits angiotensin II-induced release of aldosterone in vitro. Orally active.
Specifications
Specifications
Valsartan
>99
10mg
Valsartan
435.52