missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Valsartan
High affinity, selective AT1 antagonist
POA
Specifications
Purity | >99% |
---|---|
Gene Alias | CGP 48933 |
Product Type | Valsartan |
Molecular Weight (g/mol) | 435.52 |
Inhibitors | Valsartan |
Description
High affinity AT1 receptor antagonist (Ki = 2.38 nM). Displays 30,000-fold selectivity over AT2 receptors. Inhibits angiotensin II-induced release of aldosterone in vitro. Orally active.Specifications
>99% | |
Valsartan | |
Valsartan |
CGP 48933 | |
435.52 |
For Research Use Only