missing translation for 'onlineSavingsMsg'
Learn More

Valerophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-211690
14.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1009-14-9
C11H14O
162.23
MFCD00009480
XKGLSKVNOSHTAD-UHFFFAOYSA-N
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
66093
CHEBI:36812
1-phenylpentan-1-one
CCCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1009-14-9
C11H14O
162.23
MFCD00009480
XKGLSKVNOSHTAD-UHFFFAOYSA-N
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
66093
CHEBI:36812
1-phenylpentan-1-one
CCCCC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -9.0°C
Color Green-Yellow to Yellow
Density 0.9800g/mL
Boiling Point 105.0°C to 107.0°C (5.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CO(CH2)3CH3
Refractive Index 1.5143 to 1.5163
Quantity 5 g
Beilstein 07,327
Specific Gravity 0.98
Formula Weight 162.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valerophenone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'