Valeric anhydride, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 422450050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 2082-59-9 Weight: 0.05500kg Transport: UN number : 3265 Chem class : 1a Pack group : III


Packaging Glass bottle
Quantity 5g

Product Code. 10789911

Quantity Price
1 £ 15.4 / 5g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Valeric anhydride
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H18O3
Linear Formula (CH3(CH2)3CO)2O
Formula Weight 186.25
MDL Number MFCD00009477

Specification

Assay ≥97.5%
Physical Form Liquid
Formula Weight 186.25
Quantity 5g
Color Clear colorless to yellow
Linear Formula (CH3(CH2)3CO)2O
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 2082-59-9
Assay Percent Range 98%
Refractive Index 1.4200 to 1.4220 (20°C, 589nm)
MDL Number MFCD00009477
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C10H18O3
Density 0.9200g/mL
Chemical Name or Material Valeric anhydride
Beilstein 02, IV, 874
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.