Learn More
Gardner Denver Welch™ Kit 10 Vacuum pump control package with MRV 100, DN 25 KF
For rotary vane pumps with KF25 flanges. Cost-effective package for controlling vacuum pressure up to 0.001mbar.
Brand: Gardner Denver Welch™ 330054
Code : A0
Additional Details : Weight : 3.00000kg
Description
Vacuum control package consists of:
- MRV100 vacuum controller
- PIZA111cr-GOLD vacuum sensor
- KF25/16 measuring T-piece
- 2-way solenoid valve
- Clamps, centring rings and adapters
- Electrical connections
Specifications
DN 25 KF | |
Rotary vane pumps with KF25 flanges | |
Vacuum Control Kit |