Learn More
Vacuum Aspiration Advantage System Accessory, Single-channel Adapter
Designed for use with Fisherbrand™ Vacuum Aspiration Advantage Systems.
Brand:  Fisherbrand
Code : NEW
Additional Details : Weight : 0.15000kg
Product Code. 15441934
  • £54.75 / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Single channel plastic adapter for disposal tips with ejector
  • For use with Vacuum Aspiration Advantage Systems
Fisherbrand Aspiration Advantage System is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
Fluids can be removed easily and very precisely from slides, petri dishes, microplates, cell culture containers or similar vessels by using different pipettes or glass tips which can be easily connected to the HandVac pipettor attachment.
Specifications
Specifications
Aspiration Advantage System
Single-channel Adapter