Learn More
Fisherbrand™ Vacuum Aspiration Advantage System Accessory, Receiving Bottle
Designed for use with Fisherbrand™ Vacuum Aspiration Advantage Systems.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 1.30000kg
Description
- 4L polypropylene receiving bottle
- For Fisherbrand Aspiration Advantage System
- Includes lid, in-line filter, tubing and rapid action coupling hose connectors
Fluids can be removed easily and very precisely from slides, petri dishes, microplates, cell culture containers or similar vessels by using different pipettes or glass tips which can be easily connected to the HandVac pipettor attachment.
Specifications
Aspiration Advantage System | |
Receiving Bottle |