Learn More
BD Vacutainer™ Flashback Blood Collection Needles
Provides instant confirmation of needle placement through a visual flashback system. BD Vacutainer™ Flashback Blood Collection Needles are sterile, multi-sample needles with a safety valve and pre-attached BD tube holder. Available in several lengths and gauges.
Brand:  BD 301748
Code : 02
Additional Details : Weight : 3.50000kg
 View more versions of this product
Product Code. 12947726
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Provides instant confirmation of needle placement through a unique visual flashback indication
  • Avoids the need to probe for a vein or to stick a patient multiple times when the vein is hard to find
  • Ensures easy training of personnel and full utilization of closed system collection
  • Multi-sample needle ideally complements the range of Vacutainer blood collection line
Specifications
Specifications
Blood Collection Needle
Black
Blood Collection
22 G
31.75 mm
Sterile
Vacutainer
Complements the entire Vacutainer range of blood collection devices and accessories
1.25 in.