missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V gamma 2, PerCP-eFluor™ 710, clone: UC3-10A6, eBioscience™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 46-5828-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The UC3-10A6 monoclonal antibody reacts with the mouse V gamma 2 (Vg2) delta chain of the gamma delta T cell receptor (TCR). The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood. Studies suggest that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulating the immune response during bacterial infection.Applications Reported: This UC3-10A6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This UC3-10A6 antibody has been tested by flow cytometric analysis of C57/Bl splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
V gamma 2 TCR | |
Monoclonal | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
TCRV gamma 2, TCRV y2, TCRV gamma2, TCR V gamma2, Vg2, Vgamma2 | |
IgG | |
25μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
Murine |
Flow Cytometry | |
UC3-10A6 | |
PerCP-eFluor 710 | |
Conjugated | |
Armenian Hamster | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal |
For Research Use Only.