Learn More
Cytiva V-Bottom 96-Well Collection Plate
Convenient collection of samples in 96-well format
Brand: Cytiva 28-4039-43
Code : GE
Additional Details : Weight : 2.31790kg
Description
- Excellent fit to prepacked MultiTrap™ plates and PreDictor™ plates
- V-bottom for minimal loss of sample
- Collection of <500μL volumes of sample per well
Specifications
V-botto m collection plate | |
Polypropylene | |
Collection of eluted samples in parallel from screening plates such as MultiTrap and PreDicto r plates |
5 Pack | |
Column Accesory |