missing translation for 'onlineSavingsMsg'
Learn More

TCR V beta 6, APC, clone: RR4-7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5795-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300760

  • £143.78 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This RR4-7 monoclonal antibody reacts with mouse V beta 6 TCR. Composed of an a and b chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR4-7 antibody recognizes the V beta 6 chain on T cells from mouse strains with the a (e.g., C57BR and SJL) and b haplotypes (e.g., BALB/c, C3H, and DBA/1). Reports indicate that strains expressing the Mls-1a antigen, particularly in the presence of MHC Class II I-E, possess fewer V beta 6 TCR+ T cells.The RR4-7 antibody has been reported to have functional activity in vitro and in vivo.Applications Reported: This RR4-7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RR4-7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

V beta 6 TCR
RR4-7
APC
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Vb6-TCR, Vb-6
IgG2b
RUO
Primary
Murine
Videos