missing translation for 'onlineSavingsMsg'
Learn More

TCR V beta 6, APC, clone: RR4-7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5795-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15390750

  • £62.63 / 25µg


Estimated Shipment
06-07-2018
Add to basket

Description

Description

Description: This RR4-7 monoclonal antibody reacts with mouse V beta 6 TCR. Composed of an a and b chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR4-7 antibody recognizes the V beta 6 chain on T cells from mouse strains with the a (e.g., C57BR and SJL) and b haplotypes (e.g., BALB/c, C3H, and DBA/1). Reports indicate that strains expressing the Mls-1a antigen, particularly in the presence of MHC Class II I-E, possess fewer V beta 6 TCR+ T cells.The RR4-7 antibody has been reported to have functional activity in vitro and in vivo.Applications Reported: This RR4-7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RR4-7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

V beta 6 TCR
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
TCRV beta 6, TCRV beta6, TCR V beta6, Vb6, Vbeta6
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Murine
Flow Cytometry
RR4-7
APC
Conjugated
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.