Learn More
TCR V beta 5.1/5.2 Monoclonal Antibody (MR9-4), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£99.75 - £295.00
Specifications
Antigen | TCR V beta 5.1/5.2 |
---|---|
Clone | MR9-4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15301360
|
Affymetrix eBioscience
46-5796-82 |
100 μg |
£295.00
100µg |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15391350
|
Affymetrix eBioscience
46-5796-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This MR9-4 monoclonal antibody reacts with the mouse T cell receptor (TCR) V beta 5.1/5.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR9-4 antibody recognizes the V beta 5.1 and 5.2 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C58/J. V beta 5.1 and 5.2 are deleted in mouse strains that express MHC Class II I-E, including Balb/c, C3H, and DBA/2. Reports also indicate that V beta 5+ T cells recognize the OVA peptide. Applications Reported: This MR9-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR9-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
TCR V beta 5.1/5.2 Monoclonal antibody specifically detects TCR V beta 5.1/5.2 in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR V beta 5.1/5.2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MR9-4 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Mouse | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.