missing translation for 'onlineSavingsMsg'
Learn More

TCR V beta 2, FITC, clone: B20.6, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5804-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258259

  • £62.63 / 25µg


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The B20.6 monoclonal antibody reacts with the mouse T-cell receptor (TCR) V beta 2 chain. Neonatal T cells which express V beta 2, and are exposed to the Balb/cV virus are eliminated during negative selection, while adult T cells expressing V beta 2 and exposed to the Balb/cV virus are clonally expanded.Applications Reported: This B20.6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This B20.6 antibody has been tested by flow cytometric analysis of lymph node cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

V beta 2 TCR
B20.6
FITC
Conjugated
IgG2a
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos