Learn More
TCR V beta 13 Monoclonal Antibody (MR12-3), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 17-5797-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This MR12-3 monoclonal antibody reacts with the T cell receptor (TCR) V beta 13 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR12-3 antibody recognizes the V beta 13 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C3H. V beta 13+ T cells are deleted in mouse strains with the a (e.g., SJL) and c (e.g., RIIIS/J) haplotypes. The MR12-3 monoclonal antibody has been reported to have blocking activity. Applications Reported: This MR12-3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR12-3 antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
TCR V beta 13 Monoclonal antibody specifically detects TCR V beta 13 in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR V beta 13 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MR12-3 | |
APC | |
TCR V beta13; TCRV beta 13; TCRV beta13; Vb13; Vbeta13 | |
Affinity chromatography | |
RUO | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.