missing translation for 'onlineSavingsMsg'
Learn More

V beta 13.2 TCR Mouse anti-Human, PE, Clone: H132, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5802-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15537026

  • £66.65 / 25 tests


Estimated Shipment date
from Supplier 02-08-2017
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

This H132 monoclonal antibody recognizes the human T cell receptor (TCR) Vβ13.2 allele. Composed of an α and β chain, TCR specificity is typically determined by Vα, Jα, Vβ, Dβ, and Jβ gene rearrangement. Vβ expression in humans has been examined in studies on the effects of superantigens, inflammation, autoimmune disease, and HIV infection. More recently, assessment of TCR Vβ expression has been used to phenotype T cell clonality in CD3+/TCRαβ+ large granular lymphocyte leukemias. A member of the Ig superfamily, this receptor is expressed on a subset of peripheral blood T cells.

The H132 monoclonal antibody recognizes TCR Vβ13.2 specifically, but not Vβ13.1 or Vβ13.3.
Description & Specifications

Specifications

V beta 13.2 TCR
H132
PE
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
Conjugated
Vbeta 13.2, Vbeta13.2, Vb13.2
IgG1
RUO
Primary
Human