V beta 11 TCR Rat anti-Mouse, APC, Clone: RR3-15, eBioscience™

Rat Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 17-5827-80

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15548136

Quantity Price
1 £ 72.77 / 25µg
Estimated Shipment date
from Supplier 03-04-2017
Add to basket


Description and Specification

Specification

Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL
Gene Alias Vb11, Vbeta11
Quantity 25μg
Primary or Secondary Primary
Conjugate APC
Antigen V beta 11 TCR
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Host Species Rat
Species Reactivity Mouse
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Format Conjugated
Clone RR3-15
Applications Flow Cytometry
Regulatory Status RUO

This RR3-15 monoclonal antibody reacts with the mouse T-cell receptor (TCR) V beta 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR3-15 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b haplotype in the Tcrb gene complex, including C57BL/6 and B10, but is absent in mice with the a haplotype, such as C57L, SJL, and SWR. In addition, reports indicate that Vbeta11+ T cells are eliminated in mice bearing the MHC Class II I-E molecule. Finally, V beta 11+CD4+ T cells have been associated with graft-versus-host disease (GVHD) severity.

The RR3-15 antibody has been reported to activate V beta 11+ T cells.