missing translation for 'onlineSavingsMsg'
Learn More

V alpha 3.2 TCR Rat anti-Mouse, FITC, Clone: RR3-16, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5799-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15566566

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This RR3-16 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 3.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR3-16 antibody recognizes the V alpha 3.2 chain on T cells from mouse strains bearing the b (e.g., C57BL/6) or c haplotype (e.g., SJL, SWR, and NOD) in the Tcra gene complex. The V alpha 3.2 chain is absent in mice with the a (e.g., Balb/c, AKR, C3H) and d (e.g., DBA/1 and DBA/2) haplotypes. Studies demonstrate that the V alpha 3.2 TCR is more highly expressed on CD8+ T cells.
Specifications

Specifications

V alpha 3.2 TCR
RR3-16
FITC
Conjugated
IgG2b, kappa
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos
Documents

For Research Use Only.