missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 24 J alpha 18, PE, clone: 6B11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5806-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15557026

  • £68.96 / 25 tests

Estimated Shipment date
from Supplier 01-05-2018
Add to basket



Description: The 6B11 monoclonal antibody reacts with an epitope of the CDR3 formed by the germ-line configuration of the V alpha24 and J alpha18 of the TCR alpha locus, which results in the expression of an invariant T-cell receptor (TCR) expressed by invariant NK T cells (iNKT). This TCR is homologous to the murine V alpha 14 J alpha 18. This TCR alpha chain pairs with a restricted set of rearranged TCR beta chains, with V beta 11 being the most prominent in humans. NK T cells are restricted by the CD1d molecule and are activated by a CD1d-presented glycolipid ligand, which results in the rapid production of IL-4, IL-13 and IFN gamma. Although iNKT cells have NK-like cytolytic activity, they are considered regulators of immune responses because they rapidly produce large amounts of both Th1 and Th2 cytokines in autoimmune disease, infectious disease, and cancer. The number of iNKT cells present in peripheral blood of normal humans shows considerable variability ranging from 0.01-1% of total peripheral T cells.Applications Reported: This 6B11 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 6B11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.


V alpha 24 J alpha 18 TCR
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
TCRV alpha 24 J alpha 18, TCRV alpha24 J alpha18, TCR V alpha24 J alpha18, Valpha24, Va24
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Flow Cytometry
Affinity chromatography

For Research Use Only.