missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 24 J alpha 18, eFluor 450, clone: 6B11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5806-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15599766

  • £72.50 / 25 tests

Estimated Shipment
05-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 6B11 monoclonal antibody reacts with an epitope of the CDR3 formed by the germ-line configuration of the V alpha 24 and J alpha 18 of the TCR alpha locus, which results in the expression of an invariant T-cell receptor (TCR) expressed by invariant NK T cells (iNKT). This TCR is homologous to the murine V alpha 14J alpha 18. This TCR alpha chain pairs with a restricted set of rearranged TCR beta chains, with V beta 11 being the most prominent in humans. NK T cells are restricted by the CD1d molecule and are activated by a CD1d-presented glycolipid ligand, which results in the rapid production of IL-4, IL-13 and IFN gamma. Although iNKT cells have NK-like cytolytic activity, they are considered regulators of immune responses because they rapidly produce large amounts of both Th1 and Th2 cytokines in autoimmune disease, infectious disease, and cancer. The number of iNKT cells present in peripheral blood of normal humans shows considerable variability ranging from 0.01-1% of total peripheral T cells.Applications Reported: This 6B11 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 6B11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TCR V alpha 24 J alpha 18
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
TCRV alpha 24 J alpha 18, TCRV alpha24 J alpha18, TCR V alpha24 J alpha18, Valpha24, Va24
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Human
Flow Cytometry
6B11
eFluor 450
Liquid
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Videos
Documents

For Research Use Only.