Learn More
TCR V alpha 2 Monoclonal Antibody (B20.1), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£257.00
Specifications
Antigen | TCR V alpha 2 |
---|---|
Clone | B20.1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15508576
|
Affymetrix eBioscience
25-5812-82 |
100 μg |
£257.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The B20.1 monoclonal antibody reacts with mouse T-cell receptor (TCR) V alpha 2 chains. This antibody recognizes the majority of the TCR V alpha 2 subfamily in mice carrying the a, b and c haplotypes. The antibody also reacts with the products of T cell receptor, V delta 8 due to the high degree of homology. Applications Reported: This B20.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B20.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
TCR V alpha 2 Monoclonal antibody specifically detects TCR V alpha 2 in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR V alpha 2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG2a λ | |
Affinity chromatography | |
Antibody |
B20.1 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.