missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 2, eFluor 450, clone: B20.1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5812-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15361790

  • £62.76 / 25µg


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Description

Description

Description: The B20.1 monoclonal antibody reacts with mouse T-cell receptor (TCR) V alpha 2 chains. This antibody recognizes the majority of the TCR V alpha 2 subfamily in mice carrying the a, b and c haplotypes. The antibody also reacts with the products of T cell receptor, V delta 8 due to the high degree of homology.Applications Reported: This B20.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This B20.1 antibody has been tested. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

V alpha 2 TCR
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
TCRVA, TCRVA2, V alpha 2, V alpha-2 TCR, Valpha 2, Valpha2, Va 2, Va2
IgG2a
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Murine
Flow Cytometry
B20.1
eFluor 450
Conjugated
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.