Learn More
V alpha 11 TCR Rat anti-Mouse, PerCP-eFluor 710, Clone: RR8-1, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 46-5800-80
Code : NEW
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 15549276
  • £75.30 / 25µg
Estimated Shipment date
from Supplier 02-07-2018
Add to basket
Description
Description
This RR8-1 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR8-1 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b (e.g., C57BL/6, B10) and d (e.g., DBA/1, DBA/2) haplotypes. V alpha 11+ T cells are absent in strains with the a (e.g., BALB/c, C3H) or c (e.g., SJL) haplotypes of the Tcra gene complex. Finally, V alpha 11 is expressed predominantly on CD4+ T cells.
The RR8-1 antibody recognizes the V alpha 11.1 and 11.2 alleles, but not 11.3. This antibody has been reported to stimulate V alpha 11+ T cells.
Specifications
Specifications
V alpha 11 TCR
RR8-1
PerCP-eFluor 710
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Valpha11, Va11
IgG2b, kappa
RUO
Primary
Murine